4,6-Dichloro-5-nitropyrimidine ≥98%
Supplier: Thermo Fisher Scientific
Warning
H56252.03 H56252.06 H56252.14
H56252.03EA 70.2 EUR
H56252.03 H56252.06 H56252.14
4,6-Dichloro-5-nitropyrimidine ≥98%
4,6-Dichloro-5-nitropyrimidine
Formula: C₄HCl₂N₃O₂
MW: 193.98 g/mol
Melting Pt: 100…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00006107
CAS Number: 4316-93-2
EINECS: 224-340-6