Ampicillin sodium salt USP, Ultra Pure Grade
Supplier: VWR Chemicals
0339-EU-25GEA
228
EUR
0339-EU-25G
Ampicillin sodium salt USP, Ultra Pure Grade
Ampicillin sodium salt
Interferes with formation of bacterial cell wall. Working concentration: 50 μg/ml.
Formula:
C₁₆H₁₈N₃NaO₄S MW: 371,39 g/mol |
MDL Number:
MFCD00064313 CAS Number: 69-52-3 |
Specification Test Results
Crystallinity | PASS |
Dimethylaniline (P/F) | PASS |
Identification | PASS |
Methylene Chloride | <= 0.2 % |
pH (1%, Water) @25C | 8.0 - 10.0 |
Potency (anhydrous) | 845 - 988 mcg/mg |
Potency (as is) | 916.54 mcg/mg |
Water (KF) | <= 2.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...