5-(Aminomethyl)-2-chloropyridine 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(6-Chloropyridin-3-yl)methanamine
L19283.06
L19283.03
L19283.14
L19283.06EA
213
EUR
L19283.06
L19283.03
L19283.14
5-(Aminomethyl)-2-chloropyridine 97%
5-(Aminomethyl)-2-chloropyridine
Purity: 97%
CAS No.: 97004-04-1
Molecular Formula: C6H7ClN2
Formula Weight: 142.59
Melting Point: 27-32°
Boiling Point: 101-102°/1mm
UN No.: 3259
MDL No.: MFCD00673153
Formula:
C₆H₇ClN₂ MW: 142.59 g/mol Boiling Pt: 101…102 °C (1 mmHg) Melting Pt: 27…32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00673153 CAS Number: 97004-04-1 ADR: 8,III |