1,3-Diisopropylimidazolium tetrafluoroborate, Sigma-Aldrich®
Supplier: Merck
Synonyms: IiPr.HBF4, 1,3-Diisopropyl-1H-imidazol-3-ium tetrafluoroborate
M-Clarity™ quality level = MQ100
660019-1G 660019-5G
SIAL660019-1GEA 157 EUR
SIAL660019-1G SIAL660019-5G
1,3-Diisopropylimidazolium tetrafluoroborate, Sigma-Aldrich®
1,3-Diisopropylimidazolium tetrafluoroborate
Formula: C₉H₁₇BF₄N₂
MW: 240.0493 g/mol
MDL Number: MFCD08276804
CAS Number: 286014-34-4