Ethyl-4-chloroacetoacetate 98%
Supplier: Thermo Scientific
Danger
Synonyms: Ethyl 4-chloro-3-oxobutanoate
177640500 177642500
ACRO177640500EA 28.4 EUR
ACRO177640500 ACRO177642500
Ethyl-4-chloroacetoacetate 98%
Ethyl 4-chloroacetoacetate
Formula: C₆H₉ClO₃
MW: 164.59 g/mol
Boiling Pt: 103 °C (16 mmHg)
Melting Pt: –8 °C
Density: 1.21 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00000939
CAS Number: 638-07-3
EINECS: 211-317-0
UN: 2927
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4515 to 1.4545 (20°C, 589 nm)
GC ≥97.5 %