(3,5-Dichlorophenoxy)acetonitrile

Supplier: Thermo Scientific

SPB01308.1 SPB01308.10
MAYBSPB01308.1EA 0 EUR
MAYBSPB01308.1 MAYBSPB01308.10
(3,5-Dichlorophenoxy)acetonitrile
(3,5-Dichlorophenoxy)acetonitrile
Formula: C₈H₅Cl₂NO
MW: 202.04 g/mol
MDL Number: MFCD00052707
CAS Number: 103140-12-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR