5-Fluoro-5'-deoxyuridine (Doxifluridine), Sigma-Aldrich®
Supplier: Merck
Synonyms: (+)-5-Fluoro-5'-deoxyuridine, 1-(5-Deoxy-beta-D-ribofuranosyl)-5-fluorouracil, Doxifluridine, 5'-Deoxy-5-fluorouridine, 5'-dFUrd, 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-methyltetrahydrofuran-2-yl)-5-fluoropyrimidine-2,4(1H,3H)-dione
M-Clarity™ quality level = MQ200
F8791-100MG F8791-250MG F8791-25MG
SIALF8791-100MGEA 228 EUR
SIALF8791-100MG SIALF8791-250MG SIALF8791-25MG
5-Fluoro-5'-deoxyuridine (Doxifluridine), Sigma-Aldrich®
5-Fluoro-5'-deoxyuridine (Doxifluridine)
Formula: C₉H₁₁FN₂O₅
MW: 246.19 g/mol
MDL Number: MFCD00866530
CAS Number: 3094-09-5
EINECS: 221-440-1