Potassium cyanide-[¹³C] (99% ¹³C ), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
297038-1G 297038-250MG
SIAL297038-1GEA 431 EUR
SIAL297038-1G SIAL297038-250MG
Potassium cyanide-[¹³C] (99% ¹³C ), Sigma-Aldrich®
Potassium cyanide-[13C]
Formula: CKN
Storage Temperature: Ambient
CAS Number: 25909-68-6