Phosphonoacetic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

284270-10G 284270-50G
SIAL284270-10GEA 110 EUR
SIAL284270-10G SIAL284270-50G
Phosphonoacetic acid, Sigma-Aldrich®
Phosphonoacetic acid
Formula: C₂H₅O₅P
MW: 140.04 g/mol
MDL Number: MFCD00004311
CAS Number: 4408-78-0
EINECS: 224-558-1
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR