4-Chloro-2,6-dimethoxypyrimidine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 6-Chloro-2,4-dimethoxypyrimidine

M-Clarity™ quality level = MQ200

C36408-25G C36408-5G
SIALC36408-25GEA 502 EUR
SIALC36408-25G SIALC36408-5G
4-Chloro-2,6-dimethoxypyrimidine, Sigma-Aldrich®
4-Chloro-2,6-dimethoxypyrimidine
Formula: C₆H₇ClN₂O₂
MW: 174.59 g/mol
MDL Number: MFCD00006068
CAS Number: 6320-15-6
EINECS: 228-669-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR