2-Trimethylsilyl-1,3-dithiane, Sigma-Aldrich®

Supplier: Merck

Synonyms: (1,3-Dithian-2-yl)trimethylsilane

220817-25G 220817-5G
SIAL220817-25GEA 440 EUR
SIAL220817-25G SIAL220817-5G
2-Trimethylsilyl-1,3-dithiane, Sigma-Aldrich®
2-Trimethylsilyl-1,3-dithiane
Formula: C₇H₁₆S₂Si
MW: 192.417 g/mol
MDL Number: MFCD00006655
CAS Number: 13411-42-2
EINECS: 236-504-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR