Cumene hydroperoxide ≥80% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Cumyl hydroperoxide
M-Clarity™ quality level = MQ200
247502-100G
247502-500G
247502-5G
SIAL247502-100GEA
37.9
EUR
SIAL247502-100G
SIAL247502-500G
SIAL247502-5G
Cumene hydroperoxide ≥80% in aqueous solution, Sigma-Aldrich®
Cumene hydroperoxide
Formula:
C₉H₁₂O₂ MW: 152.20 g/mol |
MDL Number:
MFCD00002129 CAS Number: 80-15-9 EINECS: 201-254-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...