Iminodiacetic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2,2'-Azanediyldiacetic acid

M-Clarity™ quality level = MQ200

220000-25G 220000-500G
SIAL220000-25GEA 27.7 EUR
SIAL220000-25G SIAL220000-500G
Iminodiacetic acid, Sigma-Aldrich®
Iminodiacetic acid
Formula: C₄H₇NO₄
MW: 133.10 g/mol
MDL Number: MFCD00004280
CAS Number: 142-73-4
EINECS: 205-555-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR