3-Chlorobenzo[b]thiophene-2-carboxylic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

592862-1G 592862-5G
SIAL592862-1GEA 31.3 EUR
SIAL592862-1G SIAL592862-5G
3-Chlorobenzo[b]thiophene-2-carboxylic acid, Sigma-Aldrich®
3-Chlorobenzo[b]thiophene-2-carboxylic acid
Formula: C₉H₅ClO₂S
MW: 212.65 g/mol
MDL Number: MFCD00014579
CAS Number: 21211-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR