5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)

Supplier: ROCHE


M-Clarity™ quality level = MQ100

ROCH11585002001EA 481 EUR
ROCH11585002001 ROCH1585002
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
Formula: C₁₅H₁₅BrClN₂O₄P
MW: 433,6214 g/mol
MDL Number: MFCD00040636
CAS Number: 6578-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR