5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
Supplier: Roth Carl
Warning
Synonyms: BCIP, P-toluidine 5-bromo-4-chloro-1H-indol-3-yl phosphate
6368.1 6368.2 6368.3
ROTH6368.1EA 48.1 EUR
ROTH6368.1 ROTH6368.2 ROTH6368.3
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
Formula: C₁₅H₁₅BrClN₂O₄P
MW: 433.6214 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00040636
CAS Number: 6578-06-9
EINECS: 229-506-1