4-Methylumbelliferyl phosphate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

M8883-1G M8883-100MG M8883-250MG M8883-5G
SIALM8883-1GEA 286 EUR
SIALM8883-1G SIALM8883-100MG SIALM8883-250MG SIALM8883-5G
4-Methylumbelliferyl phosphate, Sigma-Aldrich®
4-Methylumbelliferyl phosphate
Formula: C₁₀H₉O₆P
MDL Number: MFCD00016969
CAS Number: 3368-04-5
EINECS: 222-137-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR