4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2), Sigma-Aldrich®
Supplier: Merck
Synonyms: Disodium 4-methylumbelliferyl phosphate, 4-MUP
M-Clarity™ quality level = MQ300
M8168-1G M8168-250MG
SIALM8168-1GEA 330 EUR
SIALM8168-1G SIALM8168-250MG
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2), Sigma-Aldrich®
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2)
Formula: C₁₀H₇Na₂O₆P
MW: 300.11 g/mol
MDL Number: MFCD00037614
CAS Number: 22919-26-2
EINECS: 245-325-0