Dibenzyl diisopropylphosphoramidite, Sigma-Aldrich®

Supplier: Merck

Synonyms: Dibenzyl N,N-diisopropylphosphoramidite

M-Clarity™ quality level = MQ200

416436-25ML 416436-5ML
SIAL416436-25MLEA 383 EUR
SIAL416436-25ML SIAL416436-5ML
Dibenzyl diisopropylphosphoramidite, Sigma-Aldrich®
Dibenzyl diisopropylphosphoramidite
Formula: C₂₀H₂₈NO₂P
MW: 345.43 g/mol
MDL Number: MFCD00191988
CAS Number: 108549-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR