(6-Bromo-2-pyridinyl)methanol 96%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: (6-Bromopyridin-2-yl)methanol, 2-Bromo-6-pyridinemethanol, 2-Bromo-6-(hydroxymethyl)pyridine
H56123.03 H56123.06
H56123.03EA 70 EUR
H56123.03 H56123.06
(6-Bromo-2-pyridinyl)methanol 96%
(6-Bromo-2-pyridinyl)methanol
Formula: C₆H₆BrNO
Boiling Pt: 246 °C
Melting Pt: 34…39 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD04039884
CAS Number: 33674-96-3