Rosolic acid (Aurin)
Supplier: Thermo Scientific
Warning
Synonyms: 4-(Bis(4-hydroxyphenyl)methylene)cyclohexa-2,5-dien-1-one
B21737.14 B21737.22
B21737.14EA 64.9 EUR
B21737.14 B21737.22
Rosolic acid (Aurin)
Rosolic acid (Aurin)
CAS No.: 603-45-2 Molecular Formula: C19H14O3 Formula Weight: 290.32 Beilstein Reference: 2055205 Merck Reference: 14,881 MDL No.: MFCD00001624
Formula: C₁₉H₁₄O₃
MW: 290.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001624
CAS Number: 603-45-2
EINECS: 210-041-8
Merck Index: 14,881