Nalidixic acid, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
N8878-5G
N8878-100G
N8878-25G
SIALN8878-5GEA
43.7
EUR
SIALN8878-5G
SIALN8878-100G
SIALN8878-25G
Nalidixic acid, Sigma-Aldrich®
Nalidixic acid
Formula:
C₁₂H₁₂N₂O₃ MW: 232.24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006884 CAS Number: 389-08-2 EINECS: 206-864-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...