Nalidixic acid sodium salt, Sigma-Aldrich®
Supplier: Merck
Synonyms: Sodium 1-ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate
MQ300
N4382-100G N4382-1G
SIALN4382-100GEA 655 EUR
SIALN4382-100G SIALN4382-1G
Nalidixic acid sodium salt, Sigma-Aldrich®
Nalidixic acid sodium salt
Formula: C₁₂H₁₁N₂NaO₃
MW: 254.22 g/mol
MDL Number: MFCD00064376
CAS Number: 3374-05-8
EINECS: 222-159-7