Diphenylboric acid-2-aminoethyl ester (2-APB)

Supplier: Roth Carl

Synonyms: 2-Aminoethyl diphenylborinate, 2-((Diphenylboryl)oxy)ethanamine

9920.1 9920.2
ROTH9920.1EA 0 EUR
ROTH9920.1 ROTH9920.2
Diphenylboric acid-2-aminoethyl ester (2-APB)
Diphenylboric acid 2-aminoethyl ester (2-APB)
Formula: C₁₄H₁₆BNO
MW: 225.1 g/mol
MDL Number: MFCD00014823
CAS Number: 524-95-8
EINECS: 208-366-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR