Luminol (3-Aminophthalhydrazide), Sigma-Aldrich®
Supplier: Merck
Synonyms: NSC 5064, Luminol, 5-Amino-2,3-Dihydro-1,4-Phthalazinedione, 5-Amino-2,3-dihydrophthalazine-1,4-dione
M-Clarity™ quality level = MQ200
A8511-25G A8511-5G
SIALA8511-25GEA 363 EUR
SIALA8511-25G SIALA8511-5G
Luminol (3-Aminophthalhydrazide), Sigma-Aldrich®
Luminol (3-Aminophthalhydrazide)
Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
MDL Number: MFCD00006890
CAS Number: 521-31-3
EINECS: 208-309-4