(-)-Carveol (mixture of cis- and trans-isomers), Sigma-Aldrich®
Supplier: Merck
Synonyms: p-Mentha-6,8-dien-2-ol, (5R)-5-Isopropenyl-2-methyl-2-cyclohexenol
M-Clarity™ quality level = MQ300
W224707-100G-K W224707-1KG-K W224707-5KG-K
SIALW224707-100G-KEA 102 EUR
SIALW224707-100G-K SIALW224707-1KG-K SIALW224707-5KG-K
(-)-Carveol (mixture of cis- and trans-isomers), Sigma-Aldrich®
(-)-Carveol (mixture of cis- and trans-isomers)
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
MDL Number: MFCD00869995
CAS Number: 99-48-9
EINECS: 202-757-4