HPLC columns, Acclaim™ Surfactant Plus
Supplier: Thermo Fisher Scientific
078960 078950 078951 078952 078953 078954 078955 078959 078956 082768 082767
555-7515EA 324 EUR
555-7515 554-4465 554-4466 554-4467 554-4468 554-4469 554-4470 555-7514 554-4503 555-7379 555-7378
HPLC columns, Acclaim™ Surfactant Plus
Chromatography Columns
These columns have been designed specifically for the HPLC separation of surfactants including anionic, non ionic, cationic and amphoteric used in a wide range of samples, such as customer products, pharmaceutical, food and beverages, environmental samples etc.
  • Unique polarity provides significantly improved resolution compared with conventional C18 columns
  • Designed for use with charged aerosol detectors and mass spectrometers
  • Can be used with evaporative light scattering detectors, suppressed conductivity detectors and UV/Visible detectors
Particle type: Ultra-pure, spherical silica
Bonded phase: Mixed-mode reversed phase and anion exchange
Hydrophobicity: Moderate to low
Carbon load: 5%
pH range: 2,5 to 7,5