Overalls, Spacel 2000 AP/EBJ, PE
Supplier: HONEYWELL SAFETY
4180131/2 4180131/3 4180131/5 4180131/6
DALL4180131/2EA 0 EUR
DALL4180131/2 DALL4180131/3 DALL4180131/5 DALL4180131/6
Overalls, Spacel 2000 AP/EBJ, PE
Disposable Garments
These disposable overalls made from PE offer protection from low risks in different work environments, e.g. pharmaceutical industry, industrial cleaning, industrial maintenance, sanitation, working in bad weather conditions.
  • Elasticated hood, zip fastener, ventilation valve, elasticated wrists, ankles and back
  • Excellent liquid tightness: Smooth, seamless surface for an excellent evacuation of liquids
  • Special designed hood for a just right tightness around the mask
  • Excellent arm and leg resistance ensures safety when moving
  • Ventilation valve for hot environment
  • Back-elastic, elasticated ankles and wrists
  • Blue colour for a better visibility
  • Sperian exclusive «Spaseal» welding
Thickness: 50 µm
Certifikace: EN 340, Cat. I