Pipette tips
Supplier: Biotix
614-1406EA 214 EUR
BITX63302165 614-1406 614-1405 BITX63302163 BITX2102.Y 614-1407 614-1413 BITX63302168 BITX63300533 BITX63300566 BITX2107.Y BITX63300520 BITX63300532 BITX63300526 614-0087 BITX63300519 BITX63302160
Pipette tips
Pipette Tips
These pipette tips are made from virgin PP or exclusive S3 polymer.
  • Polymer tips retain virtually no fluids, allowing the user to dispense truly accurate sample volumes
  • Environmentally friendly packaging: some tips available in ESP reload systems
  • Tips are certified free of RNase, DNase, DNA and endotoxins
  • Graduated sides
The exclusive S3 polymer delivers to within 0,1% of indicated volume, eliminating sample hold-up and ensuring a more precise application. This proprietary technology is essential for quantitative assays where sample retention can hinder accurate results. Universal pipette tips are made from virgin PP and are equipped with a thin wall design that provides an optimal seal with industry leading pipettors.
Eco-friendly empty pipette tip trays are manufactured from recycled PP. The trays are autoclavable, enabling reuse for maximum sustainability. When combined with ESP reload systems, these tip trays reduce plastic waste by up to 90%. The trays provide superior stability, firm placement, and a maximum space saving design.