5-Bromo-2-hydroxypyridine, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 5-Bromo-2-pyridinol, 3-Bromo-6-hydroxypyridine, 3-Bromo-6-pyridinol, 5-Bromopyridin-2-ol

M-Clarity™ quality level = MQ100

528226-25G 528226-5G
SIAL528226-25GEA 64.6 EUR
SIAL528226-25G SIAL528226-5G
5-Bromo-2-hydroxypyridine, Sigma-Aldrich®
5-Bromo-2-hydroxypyridine
Formula: C₅H₄BrNO
MW: 174 g/mol
MDL Number: MFCD00234042
CAS Number: 13466-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR