Ethyl 4-methylbenzoylformate

Supplier: Thermo Scientific

Synonyms: Ethyl 2-(4-methylphenyl)glyoxylate, Ethyl 4-Methylbenzoylformate

SPB07803.250 SPB07803.1
MAYBSPB07803.250EA 0 EUR
MAYBSPB07803.250 MAYBSPB07803.1
Ethyl 4-methylbenzoylformate
Ethyl 4-methylbenzoylformate
Formula: C₁₁H₁₂O₃
MW: 192.2100 g/mol
MDL Number: MFCD01311169
CAS Number: 5524-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR