2,6-Dibromo-4-isopropylphenyl isocyanate
Supplier: Thermo Scientific
SB01936.1 SB01936.10 SB01936.50
MAYBSB01936.1EA 0 EUR
MAYBSB01936.1 MAYBSB01936.10 MAYBSB01936.50
2,6-Dibromo-4-isopropylphenyl isocyanate
2,6-Dibromo-4-isopropylphenyl isocyanate
Formula: C₁₀H₉Br₂NO
CAS Number: 306935-84-2