Phenylglyoxylic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Oxo-2-phenylacetic acid, Benzoylformic acid

L00648.06 L00648.14
L00648.06EA 33.9 EUR
L00648.06 L00648.14
Phenylglyoxylic acid ≥98%
Phenylglyoxylic acid

Purity: 98% CAS No.: 611-73-4 Molecular Formula: C8H6O3 Formula Weight: 150.13 Melting Point: 62-67° Boiling Point: 147-151°/12mm MDL No.: MFCD00002575

Formula: C₈H₆O₃
MW: 150.13 g/mol
Boiling Pt: 147…151 °C (12 mmHg)
Melting Pt: 62…67 °C
Density: 1.38
Storage Temperature: Ambient
MDL Number: MFCD00002575
CAS Number: 611-73-4
EINECS: 210-278-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR