Diethylbenzene (mixture of isomers), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
D91055-100ML D91055-1L
SIALD91055-100MLEA 39.4 EUR
SIALD91055-100ML SIALD91055-1L
Diethylbenzene (mixture of isomers), Sigma-Aldrich®
Diethylbenzene (mixture of isomers)
Formula: C₁₀H₁₄
MW: 134.22 g/mol
MDL Number: MFCD00792900
CAS Number: 25340-17-4
EINECS: 246-874-9
UN: 2049
ADR: 3 III