Ethyl-3-(methylthio)propionate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Ethyl 3-(methylthio)propanoate
M-Clarity™ quality level = MQ300
W334308-100G-K
W334308-1KG-K
W334308-5KG-K
SIALW334308-100G-KEA
70.7
EUR
SIALW334308-100G-K
SIALW334308-1KG-K
SIALW334308-5KG-K
Ethyl-3-(methylthio)propionate, Sigma-Aldrich®
Ethyl 3-(methylthio)propionate
Formula:
C₆H₁₂O₂S MW: 148.23 g/mol |
MDL Number:
MFCD00039900 CAS Number: 13327-56-5 EINECS: 236-370-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...