Diethyl mesoxalate (diethyl ketomalonate), Sigma-Aldrich®
Supplier: Merck
Synonyms: Diethyl 2-oxomalonate
M-Clarity™ quality level = MQ100
D97401-25G D97401-5G
SIALD97401-25GEA 161 EUR
SIALD97401-25G SIALD97401-5G
Diethyl mesoxalate (diethyl ketomalonate), Sigma-Aldrich®
Diethyl mesoxalate (Diethyl ketomalonate)
Formula: C₇H₁₀O₅
MW: 174.15 g/mol
MDL Number: MFCD00009121
CAS Number: 609-09-6
EINECS: 210-176-2