7-(Trifluoromethyl)-1,2,3,4-tetrahydroquinoline ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
1,2,3,4-Tetrahydro-7-(trifluoromethyl)quinoline
L10150.03
L10150.06
L10150.MD
L10150.03EA
154
EUR
L10150.03
L10150.06
L10150.MD
7-(Trifluoromethyl)-1,2,3,4-tetrahydroquinoline ≥97%
7-(Trifluoromethyl)-1,2,3,4-tetrahydroquinoline
Purity: 97%
CAS No.: 450-62-4
Molecular Formula: C10H10F3N
Formula Weight: 201.19
Melting Point: 30-34°
Boiling Point: 135-140°/22mm
MDL No.: MFCD00079784
Formula:
C₁₀H₁₀F₃N MW: 201.19 g/mol Boiling Pt: 135…140 °C (22 mmHg) Melting Pt: 30…34 °C Storage Temperature: Ambient |
MDL Number:
MFCD00079784 CAS Number: 450-62-4 |