(S)-(+)-3-Hydroxytetrahydrofuran 98
Supplier: Thermo Fisher Scientific
Warning
H56476.06 H56476.14
H56476.06EA 59.9 EUR
H56476.06 H56476.14
(S)-(+)-3-Hydroxytetrahydrofuran 98
(S)-(+)-3-Hydroxytetrahydrofuran
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 80 °C (15 mmHg)
Density: 1.103
Flash Pt: 79 °C (174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064327
CAS Number: 86087-23-2