2-Cyanopyrazine
Supplier: Thermo Scientific
Synonyms: Pyrazine-2-carbonitrile, 2-Pyrazinecarbonitril, Pyrazinecarbonitrile
SB01106.1 SB01106.10 SB01106.50
MAYBSB01106.1EA 0 EUR
MAYBSB01106.1 MAYBSB01106.10 MAYBSB01106.50
2-Cyanopyrazine
2-Cyanopyrazine
Formula: C₅H₃N₃
MW: 105.10 g/mol
MDL Number: MFCD00049361
CAS Number: 19847-12-2
EINECS: 243-369-5
UN: 3276
ADR: 6.1,III