5-Methylpyrazine-2-carboxylic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
347647-25G
347647-5G
SIAL347647-25GEA
154
EUR
SIAL347647-25G
SIAL347647-5G
5-Methylpyrazine-2-carboxylic acid, Sigma-Aldrich®
5-Methylpyrazine-2-carboxylic acid
Formula:
C₆H₆N₂O₂ MW: 138.13 g/mol |
MDL Number:
MFCD00068241 CAS Number: 5521-55-1 EINECS: 413-260-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...