2',7'-Dichlorofluorescein, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Synonyms: 2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one
M-Clarity™ quality level = MQ200
410217-10G 410217-1G
410217-10GEA 384 EUR
410217-10G 410217-1G
2',7'-Dichlorofluorescein, Sigma-Aldrich®
2',7'-Dichlorofluorescein
Formula: C₂₀H₁₀Cl₂O₅
MDL Number: MFCD00005047
CAS Number: 76-54-0
EINECS: 200-968-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2161f301b9cb897d40057f026e346ac02c26f7d&vid=67de540d1b4d841165e31da08059ee10d5911df1&prid=web&clid=SAPDAM