4-Amino-5,6-dichlorobenzene-1,3-disulphonamide

Supplier: Thermo Scientific

RDR03841.250 RDR03841.1
MAYBRDR03841.250EA 0 EUR
MAYBRDR03841.250 MAYBRDR03841.1
4-Amino-5,6-dichlorobenzene-1,3-disulphonamide
4-Amino-5,6-dichlorobenzene-1,3-disulphonamide
Formula: C₆H₇Cl₂N₃O₄S₂
MW: 320.1735 g/mol
MDL Number: MFCD00111296
CAS Number: 5250-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR