Ethyl-4-isocyanatobenzoate

Supplier: Thermo Scientific

Synonyms: 4-(Ethoxycarbonyl)phenyl isocyanate

AC17031DA AC17031EA AC17031.50
MAYBAC17031DAEA 0 EUR
MAYBAC17031DA MAYBAC17031EA MAYBAC17031.50
Ethyl-4-isocyanatobenzoate
Ethyl 4-isocyanatobenzoate
Formula: C₁₀H₉NO₃
MDL Number: MFCD00002027
CAS Number: 30806-83-8
EINECS: 250-344-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR