(R)-(-)-1-(1-Naphthyl)ethyl isocyanate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

220442-1G 220442-5G
SIAL220442-1GEA 94.7 EUR
SIAL220442-1G SIAL220442-5G
(R)-(-)-1-(1-Naphthyl)ethyl isocyanate, Sigma-Aldrich®
(R)-(-)-1-(1-Naphthyl)ethyl isocyanate
Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
MDL Number: MFCD00004015
CAS Number: 42340-98-7
EINECS: 255-759-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR