O-Benzylhydroxylamine hydrochloride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

B22984-100G B22984-25G B22984-5G
SIALB22984-100GEA 301 EUR
SIALB22984-100G SIALB22984-25G SIALB22984-5G
O-Benzylhydroxylamine hydrochloride, Sigma-Aldrich®
O-Benzylhydroxylamine hydrochloride
Formula: C₇H₁₀ClNO
MDL Number: MFCD00012952
CAS Number: 2687-43-6
EINECS: 220-249-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR