2-(Trifluoromethyl)cyclohexanone ≥97%
Supplier: Thermo Fisher Scientific
Warning
H33423.MD H33423.03
H33423.MDEA 48.7 EUR
H33423.MD H33423.03
2-(Trifluoromethyl)cyclohexanone ≥97%
2-(Trifluoromethyl)cyclohexanone
Formula: C₇H₉F₃O
Boiling Pt: 71…73 °C (12 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00102125
CAS Number: 56734-74-8
UN: 1224
ADR: 3,III