2-Chloro-2-cyclohexylcyclohexanone

Supplier: Thermo Scientific

XBX00336.250 XBX00336.1
MAYBXBX00336.250EA 0 EUR
MAYBXBX00336.250 MAYBXBX00336.1
2-Chloro-2-cyclohexylcyclohexanone
2-Chloro-2-cyclohexylcyclohexanone
Formula: C₁₂H₁₉ClO
CAS Number: 83303-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR