O-Ethylhydroxylamine hydrochloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: Ethoxyamine hydrochloride
M-Clarity™ quality level = MQ200
274992-1G 274992-5G
SIAL274992-1GEA 49.8 EUR
SIAL274992-1G SIAL274992-5G
O-Ethylhydroxylamine hydrochloride, Sigma-Aldrich®
O-Ethylhydroxylamine hydrochloride
Formula: C₂H₈ClNO
MW: 97.54 g/mol
MDL Number: MFCD00012956
CAS Number: 3332-29-4
EINECS: 222-060-9