Hydroxylamine-O-sulphonic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: Amino hydrogen sulphate

M-Clarity™ quality level = MQ200

213136-100G 213136-25G
SIAL213136-100GEA 369 EUR
SIAL213136-100G SIAL213136-25G
Hydroxylamine-O-sulphonic acid, Sigma-Aldrich®
Hydroxylamine-O-sulphonic acid
Formula: H₃NO₄S
MW: 113.09 g/mol
MDL Number: MFCD00011604
CAS Number: 2950-43-8
EINECS: 220-971-6
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR