2-Hydroxypyridine-N-oxide, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Pyridinol-1-oxide

56413-100G 56413-25G
SIAL56413-100GEA 247 EUR
SIAL56413-100G SIAL56413-25G
2-Hydroxypyridine-N-oxide, Sigma-Aldrich®
2-Hydroxypyridine N-oxide
Formula: C₅H₅NO₂
MW: 111.10 g/mol
MDL Number: MFCD00006195
CAS Number: 13161-30-3
EINECS: 236-100-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR